Assay Details
| Gene Symbol: CEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540496.1 | XP_011538798.1 | 4 - 5 | 804 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CEPT1| Gene Symbol: CEPT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011540496.1 | XP_011538798.1 | 4 - 5 | 804 | 100 | |
Set Membership: |
|