Assay Details
Gene Symbol: TMEM99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195386.1 | NP_001182315.1 | 1 - 2 | 100 | 143 | |
GenBank mRNA | DB459363.1 | - | 1 - 2 | 100 | 143 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17000500.hg
Additional Information:
This assay is in the 5'UTR region of the NM_001195386.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17006043.hg.1 | ENST00000301665.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17006043.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17006043.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17006043.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17006043.hg.1 | TC17000500.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR17006035.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17006035.hg.1 | TC17000500.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000496847.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156817.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156817.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156817.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156817.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156817.hg.1 | TC1700007811.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000436612.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000496847.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1700007811.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | ENST00000436612.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | ENST00000301665.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | ENST00000436612.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | ENST00000622451.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | NM_001195386.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | NM_001195387.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | NM_145274.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700156819.hg.1 | TC1700007811.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 219737 | - | - |
