| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000468445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000468445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000468445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000468445.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473301.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024629.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024629.hg.1 |
ENST00000542888.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024629.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07024629.hg.1 |
TC07001618.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000468445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000468445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000468445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000468445.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473301.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0700011842.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
ENST00000005178.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
ENST00000473796.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
ENST00000498190.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
NM_002612.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700184661.hg.1 |
TC0700011842.hg.1 |
Clariom D TC |