| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000309137.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000389447.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000564108.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000565956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000566077.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000566403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000567439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000569266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
NM_024946.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
TC1600010439.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000309137.12 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000389447.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000564108.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000565956.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000566077.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000566403.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000567439.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
ENST00000569266.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1600180101.hg.1 |
NM_024946.3 |
Refseq |