| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018891.hg.1 |
ENST00000295888.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018891.hg.1 |
ENST00000322366.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018891.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018891.hg.1 |
TC04001355.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018893.hg.1 |
ENST00000295888.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018893.hg.1 |
ENST00000322366.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018893.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018893.hg.1 |
TC04001355.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018890.hg.1 |
ENST00000295888.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018890.hg.1 |
ENST00000322366.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018890.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018890.hg.1 |
TC04001355.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018892.hg.1 |
ENST00000295888.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018892.hg.1 |
ENST00000322366.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018892.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR04018892.hg.1 |
TC04001355.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
ENST00000295888.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
ENST00000502713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
ENST00000509172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
ENST00000295888.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
ENST00000502713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
ENST00000509172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176701.hg.1 |
TC0400011220.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400176702.hg.1 |
ENST00000295888.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176702.hg.1 |
ENST00000502713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176702.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176702.hg.1 |
ENST00000295888.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176702.hg.1 |
ENST00000502713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176702.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176702.hg.1 |
TC0400011220.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400176698.hg.1 |
ENST00000295888.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176698.hg.1 |
ENST00000509172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176698.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176698.hg.1 |
ENST00000295888.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176698.hg.1 |
ENST00000509172.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176698.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176698.hg.1 |
TC0400011220.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0400176700.hg.1 |
ENST00000295888.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176700.hg.1 |
ENST00000502713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176700.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176700.hg.1 |
ENST00000295888.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176700.hg.1 |
ENST00000502713.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0400176700.hg.1 |
NM_014991.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0400176700.hg.1 |
TC0400011220.hg.1 |
Clariom D TC |