Assay Details
| Gene Symbol: NMI | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004688.2 | NP_004679.2 | 1 - 2 | 325 | 105 | |
| GenBank mRNA | AK124323.1 | - | 1 - 2 | 272 | 105 | |
| AK298754.1 | - | 1 - 2 | 313 | 105 | ||
| CB151555.1 | - | 1 - 2 | 325 | 105 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002425.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK298754.1,NM_004688.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription from RNA polymerase II promoter
inflammatory response
JAK-STAT cascade
negative regulation of interferon-alpha biosynthetic process
negative regulation of interferon-beta biosynthetic process
negative regulation of innate immune response
interferon-gamma-mediated signaling pathway
positive regulation of protein K48-linked ubiquitination
regulation of nucleic acid-templated transcription
transcription cofactor activity
protein binding
transcription from RNA polymerase II promoter
inflammatory response
JAK-STAT cascade
negative regulation of interferon-alpha biosynthetic process
negative regulation of interferon-beta biosynthetic process
negative regulation of innate immune response
interferon-gamma-mediated signaling pathway
positive regulation of protein K48-linked ubiquitination
regulation of nucleic acid-templated transcription
transcription cofactor activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02038063.hg.1 | ENST00000243346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038063.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038063.hg.1 | TC02002425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038066.hg.1 | ENST00000243346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038066.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038066.hg.1 | TC02002425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038065.hg.1 | ENST00000243346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038065.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038065.hg.1 | TC02002425.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038067.hg.1 | ENST00000243346.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038067.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02038067.hg.1 | TC02002425.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206098.hg.1 | TC0200014610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206099.hg.1 | TC0200014610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200014610.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206100.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206100.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206100.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206100.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206100.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206100.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206100.hg.1 | TC0200014610.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | ENST00000243346.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | ENST00000414946.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | ENST00000477072.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | ENST00000491771.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | NM_004688.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200206095.hg.1 | TC0200014610.hg.1 | Clariom D TC |

NMI
Genomic Map