| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038059.hg.1 |
ENST00000243346.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038059.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02038059.hg.1 |
TC02002425.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000477072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000477072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000477072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000477072.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000414946.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000477072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000477072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000477072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000477072.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200014610.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200206091.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200206091.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200206091.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200206091.hg.1 |
ENST00000243346.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200206091.hg.1 |
ENST00000491771.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200206091.hg.1 |
NM_004688.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200206091.hg.1 |
TC0200014610.hg.1 |
Clariom D TC |