Assay Details
| Gene Symbol: DCK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000788.2 | NP_000779.1 | 4 - 5 | 834 | 93 | |
| GenBank mRNA | AK303292.1 | - | 4 - 5 | 910 | 93 | |
| AK313523.1 | - | 4 - 5 | 785 | 93 | ||
| BC103764.1 | - | 4 - 5 | 745 | 6472612 | 93 | |
| BC114617.1 | - | 4 - 5 | 705 | 40032871 | 93 | |
| BT019941.1 | - | 4 - 5 | 546 | 93 | ||
| BT019942.1 | - | 4 - 5 | 546 | 93 | ||
| CD014016.1 | - | 4 - 5 | 835 | 93 | ||
| CR536527.1 | - | 4 - 5 | 546 | 93 | ||
| CR541876.1 | - | 4 - 5 | 546 | 93 | ||
| JF806429.1 | - | 4 - 5 | 586 | 93 | ||
| M60527.1 | - | 4 - 5 | 705 | 93 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04002913.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
pyrimidine nucleotide metabolic process
deoxyribonucleoside monophosphate biosynthetic process
nucleotide biosynthetic process
phosphorylation
pyrimidine nucleoside salvage
purine-containing compound salvage
deoxycytidine kinase activity
ATP binding
drug binding
nucleoside kinase activity
protein homodimerization activity
pyrimidine nucleotide metabolic process
deoxyribonucleoside monophosphate biosynthetic process
nucleotide biosynthetic process
phosphorylation
pyrimidine nucleoside salvage
purine-containing compound salvage
deoxycytidine kinase activity
ATP binding
drug binding
nucleoside kinase activity
protein homodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04005431.hg.1 | ENST00000286648.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005431.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005431.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04005431.hg.1 | TC04002913.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000509617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000509617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000509617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000509617.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000509764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000509764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | ENST00000509764.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504730.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000509617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000509617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000509617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000509617.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000509764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000509764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | ENST00000509764.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012811.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | ENST00000286648.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | ENST00000503359.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | ENST00000504952.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | NM_000788.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400188465.hg.1 | TC0400012811.hg.1 | Clariom D TC |

DCK
Genomic Map