Assay Details
| Gene Symbol: EFCAB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032437.3 | NP_115813.2 | 5 - 6 | 1017 | 101 | |
| XM_005271274.3 | XP_005271331.1 | 5 - 6 | 817 | 101 | ||
| XM_006710976.3 | XP_006711039.1 | 5 - 6 | 911 | 101 | ||
| XM_006710977.1 | XP_006711040.1 | 5 - 6 | 939 | 101 | ||
| XM_011542301.2 | XP_011540603.1 | 5 - 6 | 932 | 101 | ||
| XM_017002548.1 | XP_016858037.1 | 5 - 6 | 772 | 101 | ||
| GenBank mRNA | AB058702.1 | - | 5 - 6 | 780 | 101 | |
| AK092219.1 | - | 5 - 6 | 928 | 101 | ||
| AK292220.1 | - | 5 - 6 | 976 | 101 | ||
| AK315850.1 | - | 5 - 6 | 793 | 101 | ||
| BC015814.1 | - | 5 - 6 | 721 | 3915942 | 101 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000720.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01011426.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011426.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011426.hg.1 | TC01000720.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011427.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011427.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011427.hg.1 | TC01000720.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371086.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000480886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000480886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000480886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000480886.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008588.hg.1 | NR_002771.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160907.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160907.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160907.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160907.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160907.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160907.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160907.hg.1 | TC0100008588.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371086.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000460678.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000461039.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000480886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000480886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000480886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000480886.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008588.hg.1 | NR_002771.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | ENST00000496956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160909.hg.1 | TC0100008588.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100160908.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160908.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160908.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160908.hg.1 | ENST00000371088.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160908.hg.1 | ENST00000493605.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160908.hg.1 | NM_032437.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160908.hg.1 | TC0100008588.hg.1 | Clariom D TC |

EFCAB7
Genomic Map