Assay Details
| Gene Symbol: UBAP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163692.1 | NP_001157164.1 | 3 - 4 | 56 | ||
| XM_011521547.2 | XP_011519849.1 | 3 - 4 | 56 | |||
| GenBank mRNA | BQ183317.1 | - | 2 - 3 | 349 | 56 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001559.hg.1
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15014860.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014860.hg.1 | ENST00000559089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014860.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014860.hg.1 | TC15001559.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014858.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014858.hg.1 | ENST00000559089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014858.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014858.hg.1 | TC15001559.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014863.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014863.hg.1 | ENST00000559089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014863.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014863.hg.1 | TC15001559.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014861.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014861.hg.1 | ENST00000559089.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014861.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15014861.hg.1 | TC15001559.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500172275.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172275.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172275.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172275.hg.1 | TC1500009781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172275.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172275.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172275.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | TC1500009781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172273.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | TC1500009781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172277.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | TC1500009781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172278.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172279.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172279.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172279.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172279.hg.1 | TC1500009781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172279.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172279.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172279.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009781.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | TC1500009781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172276.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | NM_001163692.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | TC1500009781.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | ENST00000502113.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | ENST00000558802.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | ENST00000559089.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | ENST00000561387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500172270.hg.1 | NM_001163692.1 | Refseq |

UBAP1L
Genomic Map