Assay Details
| Gene Symbol: RRP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015324.3 | NP_056139.1 | 3 - 4 | 1067 | 56 | |
| XM_011519955.2 | XP_011518257.1 | 3 - 4 | 1095 | 56 | ||
| XM_017017423.1 | XP_016872912.1 | 3 - 4 | 1144 | 56 | ||
| GenBank mRNA | AB007869.1 | - | 3 - 4 | 943 | 56 | |
| BC001071.2 | - | 3 - 4 | 998 | 3505288 | 56 | |
| D87060.1 | - | 3 - 4 | 919 | 56 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11003503.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
chromatin silencing at rDNA
transcription, DNA-templated
rRNA processing
covalent chromatin modification
methylation
cellular response to glucose starvation
regulation of transcription by glucose
positive regulation of cell cycle arrest
intrinsic apoptotic signaling pathway by p53 class mediator
protein binding
S-adenosylmethionine-dependent methyltransferase activity
methylated histone binding
poly(A) RNA binding
chromatin silencing at rDNA
transcription, DNA-templated
rRNA processing
covalent chromatin modification
methylation
cellular response to glucose starvation
regulation of transcription by glucose
positive regulation of cell cycle arrest
intrinsic apoptotic signaling pathway by p53 class mediator
protein binding
S-adenosylmethionine-dependent methyltransferase activity
methylated histone binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017097.hg.1 | ENST00000254605.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017097.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017097.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11017097.hg.1 | TC11003503.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203447.hg.1 | TC1100013138.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203449.hg.1 | TC1100013138.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100203450.hg.1 | TC1100013138.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000254605.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000526352.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000530762.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000533907.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | ENST00000534343.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013138.hg.1 | NM_015324.3 | Refseq |

RRP8
Genomic Map