| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024013.hg.1 |
ENST00000250366.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024013.hg.1 |
ENST00000376832.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024013.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024013.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR19024013.hg.1 |
TC19001766.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1900012012.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207489.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207489.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207489.hg.1 |
NM_012315.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1900207489.hg.1 |
TC1900012012.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1900207489.hg.1 |
ENST00000544410.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207489.hg.1 |
ENST00000594211.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1900207489.hg.1 |
NM_012315.1 |
Refseq |