Assay Details
| Gene Symbol: ETV6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001987.4 | NP_001978.1 | 1 - 2 | 305 | 77 | |
| XM_017018990.1 | XP_016874479.1 | 1 - 2 | 558 | 77 | ||
| XM_017018991.1 | XP_016874480.1 | 1 - 2 | 215 | 77 | ||
| GenBank mRNA | AK289441.1 | - | 1 - 2 | 173 | 77 | |
| BC043399.1 | - | 1 - 2 | 305 | 6014394 | 77 | |
| U11732.1 | - | 1 - 2 | 55 | 77 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003210.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK289441.1,BC043399.1,NM_001987.4,U11732.1,XM_017018990.1,XM_017018991.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
hematopoietic stem cell proliferation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein domain specific binding
negative regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
cell differentiation
positive regulation of transcription from RNA polymerase II promoter
hematopoietic stem cell proliferation
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding
protein domain specific binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12002164.hg.1 | ENST00000396373.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002164.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002164.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002164.hg.1 | TC12003210.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000266427.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000266427.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000545027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000545027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | ENST00000545027.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002163.hg.1 | ENST00000396373.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002163.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002163.hg.1 | TC12003210.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002166.hg.1 | ENST00000396373.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002166.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002166.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12002166.hg.1 | TC12003210.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147976.hg.1 | TC1200006850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | ENST00000545027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | ENST00000545027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147980.hg.1 | TC1200006850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147977.hg.1 | TC1200006850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000266427.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000266427.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000545027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000545027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | ENST00000545027.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200006850.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147975.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147975.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147975.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147975.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147975.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147975.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147975.hg.1 | TC1200006850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200147974.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147974.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147974.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147974.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147974.hg.1 | TC1200006850.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | ENST00000396373.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | ENST00000541426.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | ENST00000544715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | NM_001987.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200147979.hg.1 | TC1200006850.hg.1 | Clariom D TC |

ETV6
Genomic Map