Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002168.hg.1 |
TC12003210.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002168.hg.1 |
ENST00000396373.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12002168.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000266427.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000266427.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000545027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000545027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
ENST00000545027.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000266427.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000266427.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000541426.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000544715.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000545027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000545027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
ENST00000545027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200006850.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147984.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147984.hg.1 |
ENST00000545027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147984.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147984.hg.1 |
ENST00000396373.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147984.hg.1 |
ENST00000545027.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200147984.hg.1 |
NM_001987.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200147984.hg.1 |
TC1200006850.hg.1 |
Clariom D TC |