Assay Details
| Gene Symbol: COL18A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130445.3 | NP_569712.2 | 2 - 3 | 204 | 160 | |
| GenBank mRNA | AF018082.1 | - | 2 - 3 | 169 | 160 | |
| BC063833.1 | - | 2 - 3 | 145 | 6181818 | 160 | |
| CN389577.1 | - | 2 - 3 | 204 | 160 | ||
More Information
Updated Mapping Information
Set Membership: |
|

COL18A1
Genomic Map