| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000457045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000457045.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007631.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007631.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR10007631.hg.1 |
TC10000672.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1000157881.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157881.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157881.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000157881.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000157881.hg.1 |
TC1000008502.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000225235.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000457045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000457045.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
ENST00000485048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000008502.hg.1 |
NM_015188.1 |
Refseq |