Assay Details
| Gene Symbol: YDJC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017964.1 | NP_001017964.1 | 1 - 2 | 207 | 101 | |
| XM_011529906.2 | XP_011528208.1 | 1 - 2 | 482 | 101 | ||
| GenBank mRNA | BC096754.1 | - | 1 - 2 | 221 | 101 | |
| BC110075.1 | - | 1 - 2 | 191 | 101 | ||
| BC110799.1 | - | 1 - 2 | 192 | 101 | ||
| BC133043.1 | - | 1 - 2 | 302 | 101 | ||
| BC133047.1 | - | 1 - 2 | 302 | 101 | ||
| BI597711.1 | - | 1 - 2 | 226 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000545.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22009844.hg.1 | ENST00000292778.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009844.hg.1 | ENST00000398873.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009844.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009844.hg.1 | TC22000545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009846.hg.1 | ENST00000292778.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009846.hg.1 | ENST00000398873.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009846.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22009846.hg.1 | TC22000545.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008099.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | TC2200008099.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157945.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | NM_001017964.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | TC2200008099.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000292778.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000398873.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000415762.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000464015.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000468686.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000473985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | ENST00000482998.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200157943.hg.1 | NM_001017964.1 | Refseq |

YDJC
Genomic Map