Assay Details
| Gene Symbol: CCDC50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174908.3 | NP_777568.1 | 10 - 11 | 1480 | 89 | |
| NM_178335.2 | NP_848018.1 | 11 - 12 | 2008 | 89 | ||
| GenBank mRNA | AJ416916.2 | - | 10 - 11 | 1480 | 89 | |
| AJ557013.1 | - | 11 - 12 | 2008 | 89 | ||
| BC065004.1 | - | 10 - 11 | 977 | 6049272 | 89 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001052.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03018580.hg.1 | ENST00000392455.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018580.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018580.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018580.hg.1 | TC03001052.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018578.hg.1 | ENST00000392455.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018578.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018578.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018578.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03018578.hg.1 | TC03001052.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000460064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000460064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | ENST00000460064.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171779.hg.1 | TC0300009866.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000392456.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000460064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000460064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | ENST00000460064.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009866.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171781.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171781.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171781.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171781.hg.1 | ENST00000392455.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171781.hg.1 | NM_174908.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171781.hg.1 | NM_178335.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171781.hg.1 | TC0300009866.hg.1 | Clariom D TC |

CCDC50
Genomic Map