Assay Details
| Gene Symbol: ASAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011541755.1 | XP_011540057.1 | 9 - 10 | 892 | 56 | |
| XM_017001686.1 | XP_016857175.1 | 9 - 10 | 892 | 56 | ||
| XM_017001689.1 | XP_016857178.1 | 9 - 10 | 909 | 56 | ||
More Information
Set Membership: |
|

ASAP3
Genomic Map