Assay Details
| Gene Symbol: NUPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042483.1 | NP_001035948.1 | 1 - 2 | 380 | 70 | |
| GenBank mRNA | BF965595.1 | - | 1 - 2 | 162 | 70 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000989.hg.1
Set Membership: |
|
Gene Ontology Categories:
Function(s)
Process(es)
acute inflammatory response
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
protein complex assembly
protein acetylation
male gonad development
response to toxic substance
cell growth
positive regulation of protein modification process
skeletal muscle cell differentiation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of apoptotic process
negative regulation of cell cycle
negative regulation of fibroblast proliferation
regulation of female gonad development
molecular_function
DNA binding
chromatin binding
acute inflammatory response
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
protein complex assembly
protein acetylation
male gonad development
response to toxic substance
cell growth
positive regulation of protein modification process
skeletal muscle cell differentiation
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
positive regulation of apoptotic process
negative regulation of cell cycle
negative regulation of fibroblast proliferation
regulation of female gonad development
molecular_function
DNA binding
chromatin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR16014255.hg.1 | ENST00000324873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014255.hg.1 | ENST00000395641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014255.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014255.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014255.hg.1 | TC16000989.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014256.hg.1 | ENST00000324873.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014256.hg.1 | ENST00000395641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014256.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014256.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014256.hg.1 | TC16000989.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014257.hg.1 | ENST00000395641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014257.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014257.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014257.hg.1 | TC16000989.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014254.hg.1 | ENST00000395641.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014254.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR16014254.hg.1 | TC16000989.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1600175942.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175942.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175942.hg.1 | TC1600009865.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600175942.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175942.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175948.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175948.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175948.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175948.hg.1 | TC1600009865.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600175948.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175948.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175948.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009865.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | TC1600009865.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175944.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | TC1600009865.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | ENST00000567646.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175945.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | NM_012385.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | TC1600009865.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | ENST00000324873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | ENST00000395641.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | NM_001042483.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1600175947.hg.1 | NM_012385.2 | Refseq |

NUPR1
Genomic Map