Assay Details
| Gene Symbol: ANKRD27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032139.2 | NP_115515.2 | 21 - 22 | 2210 | 64 | |
| XM_006723412.2 | XP_006723475.1 | 21 - 22 | 2103 | 64 | ||
| XM_017027349.1 | XP_016882838.1 | 20 - 21 | 2072 | 64 | ||
| GenBank mRNA | AK054561.1 | - | 21 - 22 | 2199 | 64 | |
| AL136784.1 | - | 21 - 22 | 2187 | 64 | ||
| AL834335.1 | - | 2 - 3 | 127 | 64 | ||
| BC045605.1 | - | 21 - 22 | 2215 | 64 | ||
| BC050529.1 | - | 21 - 22 | 2166 | 6067580 | 64 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001388.hg.1
Set Membership: |
|

ANKRD27
Genomic Map