| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001672.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001672.hg.1 |
NR_003129.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_qbl_hap6001672.hg.1 |
TC6_qbl_hap6000078.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004605.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004605.hg.1 |
ENST00000427134.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004605.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004605.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004605.hg.1 |
NR_003129.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06004605.hg.1 |
TC06000395.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001303.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001303.hg.1 |
NR_003129.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mann_hap4001303.hg.1 |
TC6_mann_hap4000071.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001483.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001483.hg.1 |
NR_003129.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_mcf_hap5001483.hg.1 |
TC6_mcf_hap5000073.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001536.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001536.hg.1 |
NR_003129.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_ssto_hap7001536.hg.1 |
TC6_ssto_hap7000073.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000630.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000630.hg.1 |
NR_003129.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_apd_hap1000630.hg.1 |
TC6_apd_hap1000044.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001711.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001711.hg.1 |
NR_003129.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR6_cox_hap2001711.hg.1 |
TC6_cox_hap2000087.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600007636.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152788.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600152788.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600152788.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152788.hg.1 |
ENST00000375094.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600152788.hg.1 |
ENST00000487940.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600152788.hg.1 |
NM_006913.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600152788.hg.1 |
TC0600007636.hg.1 |
Clariom D TC |