| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000559194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
ENST00000559194.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15004088.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15004088.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15004088.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15004088.hg.1 |
TC15000437.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000557813.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000559194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
ENST00000559194.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500007346.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152363.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500152363.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152363.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152363.hg.1 |
ENST00000281282.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500152363.hg.1 |
NM_001252335.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152363.hg.1 |
NM_032866.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500152363.hg.1 |
TC1500007346.hg.1 |
Clariom D TC |