Assay Details
| Gene Symbol: CGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252335.1 | NP_001239264.1 | 19 - 20 | 3968 | 84 | |
| NM_032866.4 | NP_116255.2 | 18 - 19 | 3853 | 84 | ||
| XM_017022686.1 | XP_016878175.1 | 18 - 19 | 4486 | 84 | ||
| GenBank mRNA | AB051536.2 | - | 17 - 18 | 3012 | 84 | |
| AK027863.1 | - | 13 - 14 | 1763 | 84 | ||
| AY274808.1 | - | 18 - 19 | 3851 | 84 | ||
| AY610514.1 | - | 19 - 20 | 3948 | 84 | ||
| BC093827.1 | - | 17 - 18 | 3795 | 7939672 | 84 | |
| BC112049.1 | - | 17 - 18 | 3807 | 8327517 | 84 | |
| BC118918.1 | - | 17 - 18 | 3794 | 40074275 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000437.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15004093.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004093.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004093.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004093.hg.1 | TC15000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004094.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004094.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004094.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004094.hg.1 | TC15000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004092.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004092.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004092.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004092.hg.1 | TC15000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004090.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004090.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004090.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004090.hg.1 | TC15000437.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1500152368.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152368.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152368.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152368.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152368.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152368.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152368.hg.1 | TC1500007346.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500152366.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152366.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152366.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152366.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152366.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152366.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152366.hg.1 | TC1500007346.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500152369.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152369.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152369.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152369.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152369.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152369.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152369.hg.1 | TC1500007346.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152365.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152365.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152365.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152365.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152365.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152365.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152365.hg.1 | TC1500007346.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 106427 | - | - |

CGNL1
Genomic Map