Assay Details
| Gene Symbol: CGNL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032866.4 | NP_116255.2 | 1 - 2 | 63 | 59 | |
| XM_005254726.3 | XP_005254783.1 | 1 - 2 | 81 | 59 | ||
| GenBank mRNA | AK056673.1 | - | 1 - 2 | 63 | 59 | |
| AK307410.1 | - | 1 - 2 | 63 | 59 | ||
| AY274808.1 | - | 1 - 2 | 61 | 59 | ||
| BC030995.1 | - | 1 - 2 | 61 | 59 | ||
| DC415862.1 | - | 1 - 2 | 60 | |||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000437.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK056673.1,AY274808.1,BC030995.1,NM_032866.4,XM_005254726.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15004073.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004073.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004073.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004073.hg.1 | TC15000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004070.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004070.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004070.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15004070.hg.1 | TC15000437.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152324.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152324.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152324.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152324.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152324.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152324.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152324.hg.1 | TC1500007346.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500152344.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152344.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152344.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152344.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500152344.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152344.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500152344.hg.1 | TC1500007346.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000281282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000557813.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | ENST00000559194.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_001252335.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500007346.hg.1 | NM_032866.4 | Refseq |

CGNL1
Genomic Map