Assay Details
| Gene Symbol: BRAF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004333.4 | NP_004324.2 | 7 - 8 | 1033 | 81 | |
| XM_005250045.2 | XP_005250102.1 | 7 - 8 | 980 | 81 | ||
| XM_017012558.1 | XP_016868047.1 | 7 - 8 | 980 | 81 | ||
| XM_017012559.1 | XP_016868048.1 | 7 - 8 | 980 | 81 | ||
| GenBank mRNA | AK314346.1 | - | 7 - 8 | 980 | 81 | |
| BC101757.1 | - | 7 - 8 | 994 | 8069263 | 81 | |
| BC112079.1 | - | 7 - 8 | 994 | 8327547 | 81 | |
| M95712.2 | - | 7 - 8 | 1033 | 81 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07001927.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPKK activity
myeloid progenitor cell differentiation
protein phosphorylation
visual learning
organ morphogenesis
positive regulation of gene expression
negative regulation of fibroblast migration
glucose transport
thyroid gland development
positive regulation of peptidyl-serine phosphorylation
somatic stem cell population maintenance
cellular response to drug
regulation of cell proliferation
negative regulation of apoptotic process
CD4-positive, alpha-beta T cell differentiation
positive T cell selection
response to peptide hormone
negative regulation of neuron apoptotic process
thymus development
positive regulation of axon regeneration
positive regulation of axonogenesis
protein heterooligomerization
positive regulation of stress fiber assembly
response to cAMP
long-term synaptic potentiation
head morphogenesis
face development
positive regulation of ERK1 and ERK2 cascade
cellular response to calcium ion
establishment of protein localization to membrane
positive regulation of substrate adhesion-dependent cell spreading
negative regulation of synaptic vesicle exocytosis
negative regulation of endothelial cell apoptotic process
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
calcium ion binding
protein binding
ATP binding
small GTPase binding
mitogen-activated protein kinase kinase binding
identical protein binding
protein heterodimerization activity
MAPK cascade
activation of MAPKK activity
myeloid progenitor cell differentiation
protein phosphorylation
visual learning
organ morphogenesis
positive regulation of gene expression
negative regulation of fibroblast migration
glucose transport
thyroid gland development
positive regulation of peptidyl-serine phosphorylation
somatic stem cell population maintenance
cellular response to drug
regulation of cell proliferation
negative regulation of apoptotic process
CD4-positive, alpha-beta T cell differentiation
positive T cell selection
response to peptide hormone
negative regulation of neuron apoptotic process
thymus development
positive regulation of axon regeneration
positive regulation of axonogenesis
protein heterooligomerization
positive regulation of stress fiber assembly
response to cAMP
long-term synaptic potentiation
head morphogenesis
face development
positive regulation of ERK1 and ERK2 cascade
cellular response to calcium ion
establishment of protein localization to membrane
positive regulation of substrate adhesion-dependent cell spreading
negative regulation of synaptic vesicle exocytosis
negative regulation of endothelial cell apoptotic process
protein kinase activity
protein serine/threonine kinase activity
MAP kinase kinase kinase activity
calcium ion binding
protein binding
ATP binding
small GTPase binding
mitogen-activated protein kinase kinase binding
identical protein binding
protein heterodimerization activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07029952.hg.1 | TC07001927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029952.hg.1 | ENST00000288602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029952.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029953.hg.1 | TC07001927.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029953.hg.1 | ENST00000288602.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07029953.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000469930.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000479537.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013629.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700202221.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700202221.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700202221.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700202221.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700202221.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700202221.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700202221.hg.1 | TC0700013629.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700202219.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700202219.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700202219.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700202219.hg.1 | ENST00000288602.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700202219.hg.1 | ENST00000497784.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700202219.hg.1 | NM_004333.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700202219.hg.1 | TC0700013629.hg.1 | Clariom D TC |

BRAF
Genomic Map