Assay Details
| Gene Symbol: LOC101927359 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125885.1 | 1 - 2 | 87 | 156 | ||
| GenBank mRNA | BC040219.1 | - | 1 - 2 | 84 | 156 | |
| DA255139.1 | - | 1 - 2 | 87 | 156 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181538.hg.1 | TC0400011870.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181539.hg.1 | TC0400011870.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400181537.hg.1 | TC0400011870.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000505289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000506897.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | ENST00000509715.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011870.hg.1 | NR_125885.1 | Refseq |

LOC101927359
Genomic Map