| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008591.hg.1 |
ENST00000370435.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008591.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008591.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008591.hg.1 |
TC06000720.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008593.hg.1 |
ENST00000370435.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008593.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008593.hg.1 |
TC06000720.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008594.hg.1 |
ENST00000370435.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008594.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008594.hg.1 |
TC06000720.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008588.hg.1 |
ENST00000370435.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008588.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008588.hg.1 |
TC06000720.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008590.hg.1 |
ENST00000370435.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008590.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008590.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008590.hg.1 |
TC06000720.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008595.hg.1 |
ENST00000370435.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008595.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008595.hg.1 |
TC06000720.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008585.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR06008585.hg.1 |
TC06000720.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158990.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158990.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158990.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158990.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158990.hg.1 |
TC0600008486.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600158993.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158993.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158993.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158993.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158993.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158993.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158993.hg.1 |
TC0600008486.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0600158987.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158987.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158987.hg.1 |
TC0600008486.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
ENST00000467503.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0600008486.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158991.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158991.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158991.hg.1 |
ENST00000370435.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0600158991.hg.1 |
NM_024576.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0600158991.hg.1 |
TC0600008486.hg.1 |
Clariom D TC |