Assay Details
| Gene Symbol: P3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006455.2 | NP_006446.1 | 3 - 4 | 1073 | 79 | |
| XM_006721640.3 | XP_006721703.1 | 3 - 4 | 1491 | 79 | ||
| GenBank mRNA | AJ250583.1 | - | 3 - 4 | 874 | 79 | |
| AK056085.1 | - | 4 - 5 | 1256 | 79 | ||
| AK222945.1 | - | 3 - 4 | 818 | 79 | ||
| AK301301.1 | - | 4 - 5 | 790 | 79 | ||
| BC001047.2 | - | 3 - 4 | 826 | 3510406 | 79 | |
| BC007942.2 | - | 3 - 4 | 821 | 4130726 | 79 | |
| BC011701.2 | - | 3 - 4 | 805 | 3510378 | 79 | |
| U47621.1 | - | 3 - 4 | 800 | 79 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001517.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17020109.hg.1 | ENST00000355468.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020109.hg.1 | ENST00000393928.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020109.hg.1 | ENST00000545545.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020109.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17020109.hg.1 | TC17001517.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000465097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000465097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000465097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000465097.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000590496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000590496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000590496.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184681.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184681.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184681.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700184681.hg.1 | TC1700010687.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700184681.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184681.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700184681.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000355468.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000393928.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000465097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000465097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000465097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000465097.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000467164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000484247.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000587455.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000590496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000590496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000590496.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | ENST00000592026.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010687.hg.1 | NM_006455.2 | Refseq |

P3H4
Genomic Map