Assay Details
| Gene Symbol: ACBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032360.3 | NP_115736.1 | 3 - 4 | 1002 | 110 | |
| GenBank mRNA | AK314112.1 | - | 3 - 4 | 977 | 110 | |
| BC006505.2 | - | 3 - 4 | 459 | 2821736 | 110 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01006385.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01055362.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055362.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055362.hg.1 | TC01006385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055365.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055365.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055365.hg.1 | TC01006385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055361.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055361.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055361.hg.1 | TC01006385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055363.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055363.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055363.hg.1 | TC01006385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055364.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055364.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01055364.hg.1 | TC01006385.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244260.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244260.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244260.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244260.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244260.hg.1 | TC0100018540.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100244258.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244258.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244258.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244258.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244258.hg.1 | TC0100018540.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | ENST00000475338.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100018540.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244255.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244255.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244255.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244255.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244255.hg.1 | TC0100018540.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100244259.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244259.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244259.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244259.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244259.hg.1 | TC0100018540.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100244256.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244256.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244256.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244256.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244256.hg.1 | TC0100018540.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100244257.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244257.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244257.hg.1 | ENST00000367595.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100244257.hg.1 | NM_032360.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100244257.hg.1 | TC0100018540.hg.1 | Clariom D TC |

ACBD6
Genomic Map