Assay Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018279.3 | NP_060749.2 | 4 - 5 | 1224 | 66 | |
| GenBank mRNA | AK001798.1 | - | 4 - 5 | 826 | 66 | |
| AK223148.1 | - | 4 - 5 | 822 | 66 | ||
| AK315587.1 | - | 4 - 5 | 783 | 66 | ||
| BC008596.1 | - | 4 - 5 | 1224 | 4154303 | 66 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12003254.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12008339.hg.1 | ENST00000266673.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008339.hg.1 | ENST00000549735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008339.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008339.hg.1 | TC12003254.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008340.hg.1 | ENST00000266673.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008340.hg.1 | ENST00000549735.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008340.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12008340.hg.1 | TC12003254.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201043.hg.1 | TC1200012666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201044.hg.1 | TC1200012666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200201045.hg.1 | TC1200012666.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000266673.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546677.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000546795.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000549735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550524.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | ENST00000550787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012666.hg.1 | NM_018279.3 | Refseq |

TMEM19
Genomic Map