Assay Details
| Gene Symbol: PCGF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011663.1 | NP_001011663.1 | 8 - 9 | 971 | 102 | |
| NM_032154.3 | NP_115530.2 | 5 - 6 | 746 | 102 | ||
| GenBank mRNA | AB047006.1 | - | 8 - 9 | 925 | 102 | |
| AK027885.1 | - | 8 - 9 | 947 | 102 | ||
| AK290679.1 | - | 8 - 9 | 925 | 102 | ||
| AK309105.1 | - | 9 - 10 | 1113 | 102 | ||
| BC007602.1 | - | 5 - 6 | 697 | 3350074 | 102 | |
| BC010235.1 | - | 8 - 9 | 977 | 3461513 | 102 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001628.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
negative regulation of transcription from RNA polymerase II promoter
transcription, DNA-templated
negative regulation of transcription, DNA-templated
RNA polymerase II regulatory region sequence-specific DNA binding
transcriptional repressor activity, RNA polymerase II transcription regulatory region sequence-specific binding
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000492755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000492755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | ENST00000492755.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021266.hg.1 | ENST00000337211.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021266.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021266.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021266.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021266.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10021266.hg.1 | TC10001628.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000492755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000492755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | ENST00000492755.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000011713.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | ENST00000337211.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | ENST00000369847.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | ENST00000490296.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | NM_001011663.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | NM_032154.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000180166.hg.1 | TC1000011713.hg.1 | Clariom D TC |

PCGF6
Genomic Map