Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001281789.1 | NP_001268718.1 | 2 - 3 | 207 | 105 | |
| NM_153261.5 | NP_694993.2 | 3 - 4 | 258 | 105 | ||
| GenBank mRNA | AK095420.1 | - | 2 - 3 | 180 | 105 | |
| AK301610.1 | - | 3 - 4 | 223 | 105 | ||
| AK307132.1 | - | 2 - 3 | 254 | 105 | ||
| BC036683.2 | - | 3 - 4 | 187 | 5266084 | 105 | |
| FJ516381.1 | - | 2 - 3 | 135 | 105 | ||
| HY011229.1 | - | 2 - 3 | 208 | 105 | ||
More Information
Set Membership: |
|

CNEP1R1
Genomic Map