Assay Details
| Gene Symbol: DOCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290223.1 | NP_001277152.1 | 32 - 33 | 3395 | 97 | |
| NM_001380.4 | NP_001371.1 | 32 - 33 | 3332 | 97 | ||
| XM_011539422.2 | XP_011537724.1 | 32 - 33 | 3373 | 97 | ||
| XM_011539424.1 | XP_011537726.1 | 32 - 33 | 3283 | 97 | ||
| XM_017015813.1 | XP_016871302.1 | 32 - 33 | 3305 | 97 | ||
| XM_017015816.1 | XP_016871305.1 | 15 - 16 | 1553 | 97 | ||
| GenBank mRNA | BC146857.1 | - | 32 - 33 | 3285 | 9056785 | 97 |
| BX648342.1 | - | 4 - 5 | 1420 | 97 | ||
| D50857.1 | - | 32 - 33 | 3246 | 97 | ||
More Information
Updated Mapping Information
Set Membership: |
|

DOCK1
Genomic Map