Assay Details
| Gene Symbol: SLU7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006425.4 | NP_006416.3 | 8 - 9 | 970 | 119 | |
| XM_005265793.3 | XP_005265850.1 | 8 - 9 | 1317 | 119 | ||
| XM_011534418.2 | XP_011532720.1 | 9 - 10 | 1020 | 119 | ||
| GenBank mRNA | AF101074.1 | - | 7 - 8 | 815 | 119 | |
| AK024875.1 | - | 8 - 9 | 890 | 119 | ||
| AK314564.1 | - | 8 - 9 | 890 | 119 | ||
| BC010634.1 | - | 8 - 9 | 896 | 3871882 | 119 | |
| CA976106.1 | - | 6 - 7 | 561 | 119 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05002010.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
RNA splicing, via transesterification reactions
alternative mRNA splicing, via spliceosome
mRNA 3'-splice site recognition
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
intracellular protein transport
mRNA 3'-end processing
cellular response to heat
second spliceosomal transesterification activity
protein binding
zinc ion binding
pre-mRNA 3'-splice site binding
RNA splicing, via transesterification reactions
alternative mRNA splicing, via spliceosome
mRNA 3'-splice site recognition
mRNA splicing, via spliceosome
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
intracellular protein transport
mRNA 3'-end processing
cellular response to heat
second spliceosomal transesterification activity
protein binding
zinc ion binding
pre-mRNA 3'-splice site binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521320.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027922.hg.1 | ENST00000297151.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027922.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05027922.hg.1 | TC05002010.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000518268.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000519349.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520664.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521190.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521320.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000521826.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | ENST00000523219.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500012663.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500188378.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500188378.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500188378.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500188378.hg.1 | ENST00000297151.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500188378.hg.1 | ENST00000520841.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500188378.hg.1 | NM_006425.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500188378.hg.1 | TC0500012663.hg.1 | Clariom D TC |

SLU7
Genomic Map