| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
ENST00000275607.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
ENST00000342190.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
ENST00000395436.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
ENST00000413756.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
ENST00000434526.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
NM_001042469.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
NM_001130069.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
NM_001146333.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
NM_015411.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR07005083.hg.1 |
TC07000341.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000275607.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000342190.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000395436.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000413756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000413952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000423763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000434526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000436782.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000438133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000447501.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000451338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000452216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000461912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000531229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
NM_001042469.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
NM_001130069.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
NM_001146333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
NM_015411.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000275607.13 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000342190.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000395436.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000413756.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000413952.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000423763.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000434526.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000436782.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000438133.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000447501.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000451338.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000452216.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000461912.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
ENST00000531229.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
NM_001042469.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
NM_001130069.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
NM_001146333.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
NM_015411.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0700152904.hg.1 |
TC0700007633.hg.1 |
Clariom D TC |