Assay Details
| Gene Symbol: MOSPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001306188.1 | NP_001293117.1 | 2 - 3 | 411 | 117 | |
| NM_019556.2 | NP_062456.1 | 2 - 3 | 411 | 117 | ||
| XM_011531366.2 | XP_011529668.1 | 2 - 3 | 365 | 117 | ||
| XM_017029653.1 | XP_016885142.1 | 3 - 4 | 604 | 117 | ||
| XM_017029654.1 | XP_016885143.1 | 2 - 3 | 365 | 117 | ||
| GenBank mRNA | AK299074.1 | - | 2 - 3 | 324 | 117 | |
| AK299172.1 | - | 1 - 2 | 201 | 117 | ||
| AK316570.1 | - | 2 - 3 | 286 | 117 | ||
| AL137163.1 | - | 2 - 3 | 334 | 117 | ||
| BC005700.1 | - | 2 - 3 | 314 | 3450155 | 117 | |
| CB997365.1 | - | 2 - 3 | 319 | 117 | ||
| CN428237.1 | - | 2 - 3 | 413 | 117 | ||
| DQ323992.1 | - | 1 - 2 | 147 | 117 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC0X001397.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018619.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018619.hg.1 | ENST00000370779.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018619.hg.1 | ENST00000370783.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018619.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018619.hg.1 | TC0X001397.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018617.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018617.hg.1 | ENST00000370779.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018617.hg.1 | ENST00000370783.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018617.hg.1 | ENST00000491609.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018617.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X018617.hg.1 | TC0X001397.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00173887.hg.1 | TC0X00010880.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370777.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370779.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000370783.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000462060.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000480721.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000489890.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | ENST00000491609.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_001306188.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00010880.hg.1 | NM_019556.2 | Refseq |

MOSPD1
Genomic Map