Assay Details
| Gene Symbol: C8orf49 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103552.1 | 1 | 734 | 66 | ||
| GenBank mRNA | AK055534.1 | - | 1 | 734 | 66 | |
| BC132945.1 | - | 1 | 603 | 40146967 | 66 | |
| BC136882.1 | - | 1 | 603 | 9020873 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 949031 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 33940 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3381996 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P32206540 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8144657 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8144656 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1564315_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | ENST00000625198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | ENST00000625198.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145776.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145776.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145776.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145776.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145776.hg.1 | TC0800006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800145777.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145777.hg.1 | ENST00000625198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145777.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145777.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145777.hg.1 | ENST00000625198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145777.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145777.hg.1 | TC0800006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | ENST00000625198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | ENST00000625198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800006757.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145778.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145778.hg.1 | ENST00000625198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145778.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145778.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145778.hg.1 | ENST00000625198.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145778.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145778.hg.1 | TC0800006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800145779.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145779.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145779.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145779.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145779.hg.1 | TC0800006757.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800145780.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145780.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145780.hg.1 | ENST00000525043.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800145780.hg.1 | NR_103552.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800145780.hg.1 | TC0800006757.hg.1 | Clariom D TC |
| Human Genome U133 Plus 2.0 Array | 1564315_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1564315_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1564315_at6 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 230624 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3307154 | BC132945.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3307154 | BC136882.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3307154 | AK055534.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1774884 | AK055534.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1774884 | AK055534.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1774884 | AK055534.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1774884 | AK055534.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1774884 | AK055534.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1774884 | AK055534.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1774884 | AK055534.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1774884 | AK055534.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1774884 | AK055534.1 | human_transcriptome |

C8orf49
Genomic Map