| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000234.hg.1 |
NM_001145146.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000234.hg.1 |
NM_001145147.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000234.hg.1 |
NM_001145148.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000234.hg.1 |
NM_004382.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17_ctg5_hap1000234.hg.1 |
TC17_ctg5_hap1000028.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
ENST00000314537.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
ENST00000347197.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
ENST00000352855.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
ENST00000398285.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
ENST00000535778.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
NM_001145146.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
NM_001145147.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
NM_001145148.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
NM_004382.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR17007222.hg.1 |
TC17002876.hg.1 |
HTA TC |