Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320669.1 | NP_001307598.1 | 1 - 2 | 78 | 53 | |
| NM_001320670.1 | NP_001307599.1 | 1 - 2 | 78 | 53 | ||
| GenBank mRNA | DA802473.1 | - | 1 - 2 | 44 | 53 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001320669.1, NM_001320670.1 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |

ZFP30
Genomic Map