Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001320666.1 | NP_001307595.1 | 4 - 5 | 751 | 120 | |
| NM_001320667.1 | NP_001307596.1 | 4 - 5 | 657 | 120 | ||
| NM_001320668.1 | NP_001307597.1 | 4 - 5 | 527 | 120 | ||
| NM_001320669.1 | NP_001307598.1 | 4 - 5 | 465 | 120 | ||
| NM_014898.2 | NP_055713.1 | 4 - 5 | 698 | 120 | ||
| GenBank mRNA | AB023178.1 | - | 4 - 5 | 698 | 120 | |
| BC041087.1 | - | 4 - 5 | 727 | 5579247 | 120 | |
| BC050542.1 | - | 4 - 5 | 656 | 120 | ||
| GU388430.1 | - | 4 - 5 | 527 | 120 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19001486.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19020166.hg.1 | ENST00000351218.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19020166.hg.1 | ENST00000392144.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19020166.hg.1 | ENST00000440715.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19020166.hg.1 | ENST00000514101.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19020166.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19020166.hg.1 | TC19001486.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | TC1900011935.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205848.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | TC1900011935.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900205849.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000351218.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000392144.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000477900.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000514101.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000587809.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000588979.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589018.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000589789.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | ENST00000591444.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900011935.hg.1 | NM_014898.3 | Refseq |

ZFP30
Genomic Map