| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005500.hg.1 |
ENST00000265562.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005500.hg.1 |
ENST00000456408.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005500.hg.1 |
NM_015466.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR03005500.hg.1 |
TC03000268.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
ENST00000265562.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
ENST00000602307.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
NM_001304482.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
NM_015466.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
ENST00000265562.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
ENST00000602307.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
NM_001304482.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
NM_015466.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0300151761.hg.1 |
TC0300007294.hg.1 |
Clariom D TC |