Assay Details
| Gene Symbol: GABPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323906.1 | NP_001310835.1 | 6 - 7 | 1090 | 122 | |
| NM_001323908.1 | NP_001310837.1 | 6 - 7 | 1122 | 122 | ||
| NM_001323910.1 | NP_001310839.1 | 7 - 8 | 1138 | 122 | ||
| NM_001323913.1 | NP_001310842.1 | 5 - 6 | 922 | 122 | ||
| NM_144618.2 | NP_653219.1 | 6 - 7 | 1064 | 122 | ||
| XM_017000246.1 | XP_016855735.1 | 7 - 8 | 1568 | 122 | ||
| XM_017000247.1 | XP_016855736.1 | 8 - 9 | 1655 | 122 | ||
| XM_017000248.1 | XP_016855737.1 | 6 - 7 | 964 | 122 | ||
| XM_017000249.1 | XP_016855738.1 | 7 - 8 | 1024 | 122 | ||
| XM_017000252.1 | XP_016855741.1 | 6 - 7 | 810 | 122 | ||
| XM_017000253.1 | XP_016855742.1 | 6 - 7 | 781 | 122 | ||
| GenBank mRNA | AK092230.1 | - | 6 - 7 | 904 | 122 | |
| AK313606.1 | - | 6 - 7 | 877 | 122 | ||
| BC027033.1 | - | 6 - 7 | 1064 | 5139830 | 122 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001202.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01018563.hg.1 | TC01001202.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018563.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018563.hg.1 | ENST00000368917.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018563.hg.1 | ENST00000368918.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018563.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01018563.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000462520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000486076.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009939.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | ENST00000368916.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | ENST00000368918.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | ENST00000467551.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | ENST00000489549.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | NM_144618.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100171924.hg.1 | TC0100009939.hg.1 | Clariom D TC |

GABPB2
Genomic Map