Assay Details
Gene Symbol: NOTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017617.4 | NP_060087.3 | 34 | 8984 | 157 | |
GenBank mRNA | AB209873.1 | - | 26 | 7521 | 157 | |
AK000012.1 | - | 1 | 1160 | 157 | ||
BC013208.1 | - | 4 | 3169 | 157 | ||
BC046127.1 | - | 1 | 735 | 157 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
