Assay Details
| Gene Symbol: CCDC73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008391.3 | NP_001008392.2 | 9 - 10 | 700 | 115 | |
| XM_011520139.2 | XP_011518441.1 | 8 - 9 | 561 | 115 | ||
| GenBank mRNA | AK131470.1 | - | 9 - 10 | 683 | 115 | |
| BC062749.1 | - | 9 - 10 | 700 | 115 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11001534.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000528333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000528333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000528333.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019231.hg.1 | ENST00000335185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019231.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019231.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019231.hg.1 | TC11001534.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019232.hg.1 | ENST00000335185.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019232.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019232.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11019232.hg.1 | TC11001534.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100179433.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100179433.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100179433.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100179433.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100179433.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100179433.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100179433.hg.1 | TC1100010472.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100179434.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100179434.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100179434.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100179434.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100179434.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100179434.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100179434.hg.1 | TC1100010472.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000335185.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000528333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000528333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000528333.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000531481.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | ENST00000534415.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100010472.hg.1 | NM_001008391.3 | Refseq |

CCDC73
Genomic Map