Assay Details
| Gene Symbol: CLYBL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206808.3 | NP_996531.1 | 8 - 9 | 1071 | 100 | |
| XM_005254031.2 | XP_005254088.1 | 7 - 8 | 948 | 100 | ||
| XM_017020403.1 | XP_016875892.1 | 7 - 8 | 1769 | 100 | ||
| GenBank mRNA | AF428253.1 | - | 8 - 9 | 1039 | 100 | |
| BC034360.1 | - | 8 - 9 | 1053 | 4750050 | 100 | |
| BP313488.1 | - | 2 - 3 | 252 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000358.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13003801.hg.1 | ENST00000376355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003801.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003801.hg.1 | TC13000358.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003799.hg.1 | ENST00000376355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003799.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003799.hg.1 | TC13000358.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003797.hg.1 | ENST00000339105.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003797.hg.1 | ENST00000376355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003797.hg.1 | ENST00000376360.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003797.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003797.hg.1 | TC13000358.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003800.hg.1 | ENST00000376355.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003800.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003800.hg.1 | TC13000358.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376354.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000416504.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000419700.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000425186.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | ENST00000443887.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007845.hg.1 | NR_104592.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152826.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152826.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152826.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152826.hg.1 | ENST00000339105.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152826.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152826.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152826.hg.1 | TC1300007845.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300152828.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152828.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152828.hg.1 | ENST00000376355.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152828.hg.1 | NM_206808.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152828.hg.1 | TC1300007845.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 124662 | - | - |

CLYBL
Genomic Map