Assay Details
| Gene Symbol: MPZL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286152.1 | NP_001273081.1 | 2 - 3 | 277 | 120 | |
| NM_198275.2 | NP_938016.1 | 2 - 3 | 313 | 120 | ||
| XM_006718775.2 | XP_006718838.1 | 2 - 3 | 665 | 120 | ||
| XM_017017304.1 | XP_016872793.1 | 2 - 3 | 424 | 120 | ||
| GenBank mRNA | AK095399.1 | - | 2 - 3 | 265 | 120 | |
| AK289390.1 | - | 2 - 3 | 265 | 120 | ||
| AK304292.1 | - | 2 - 3 | 256 | 120 | ||
| AY358626.1 | - | 2 - 3 | 241 | 120 | ||
| BC113586.1 | - | 2 - 3 | 295 | 8322638 | 120 | |
| BC143897.1 | - | 2 - 3 | 259 | 9052415 | 120 | |
| DC417020.1 | - | 2 - 3 | 313 | 120 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002345.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR11028153.hg.1 | ENST00000278949.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028153.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028153.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028153.hg.1 | TC11002345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028152.hg.1 | ENST00000278949.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028152.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028152.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11028152.hg.1 | TC11002345.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000446386.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000525386.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104404.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012473.hg.1 | NR_104405.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | ENST00000278949.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | ENST00000527472.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | NM_001286152.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | NM_198275.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100195922.hg.1 | TC1100012473.hg.1 | Clariom D TC |

MPZL3
Genomic Map