| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050078.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050078.hg.1 |
ENST00000451038.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050078.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050078.hg.1 |
TC01003247.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050079.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050079.hg.1 |
ENST00000451038.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050079.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01050079.hg.1 |
TC01003247.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015832.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015832.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015832.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015832.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015832.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015832.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015832.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0100015832.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015832.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015832.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015832.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015832.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0100015832.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015832.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015832.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0100015832.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218941.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218941.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218941.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218941.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218941.hg.1 |
TC0100015832.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0100218943.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218943.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218943.hg.1 |
ENST00000271835.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100218943.hg.1 |
NM_016190.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100218943.hg.1 |
TC0100015832.hg.1 |
Clariom D TC |