Assay Details
| Gene Symbol: CRIM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016441.2 | NP_057525.1 | 3 - 4 | 816 | 83 | |
| XM_005264357.4 | XP_005264414.1 | 2 - 3 | 638 | 83 | ||
| XM_011532898.2 | XP_011531200.1 | 4 - 5 | 937 | 83 | ||
| XM_011532899.2 | XP_011531201.1 | 4 - 5 | 938 | 83 | ||
| XM_011532901.2 | XP_011531203.1 | 4 - 5 | 938 | 83 | ||
| XM_017004259.1 | XP_016859748.1 | 3 - 4 | 955 | 83 | ||
| GenBank mRNA | AB209139.1 | - | 3 - 4 | 816 | 83 | |
| AF167706.1 | - | 3 - 4 | 789 | 83 | ||
| AY358371.1 | - | 3 - 4 | 1117 | 83 | ||
| BC111989.1 | - | 3 - 4 | 787 | 8327457 | 83 | |
| BC113371.1 | - | 3 - 4 | 787 | 8322423 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000230.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cell growth
nervous system development
negative regulation of endopeptidase activity
insulin-like growth factor receptor signaling pathway
serine-type endopeptidase inhibitor activity
insulin-like growth factor-activated receptor activity
insulin-like growth factor binding
PDZ domain binding
regulation of cell growth
nervous system development
negative regulation of endopeptidase activity
insulin-like growth factor receptor signaling pathway
serine-type endopeptidase inhibitor activity
insulin-like growth factor-activated receptor activity
insulin-like growth factor binding
PDZ domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000413985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000413985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000413985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000413985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000428774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000428774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000428774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000428774.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000473403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000473403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000473403.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000477491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000477491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000477491.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000481321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000481321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000481321.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000497236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | ENST00000497236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003397.hg.1 | ENST00000280527.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003397.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02003397.hg.1 | TC02000230.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000413985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000413985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000413985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000413985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000428774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000428774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000428774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000428774.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000473403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000473403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000473403.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000477491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000477491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000477491.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000481321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000481321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000481321.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000497236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | ENST00000497236.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007237.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150088.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150088.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150088.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150088.hg.1 | ENST00000280527.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150088.hg.1 | ENST00000426856.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200150088.hg.1 | NM_016441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200150088.hg.1 | TC0200007237.hg.1 | Clariom D TC |

CRIM1
Genomic Map