| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017379.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017379.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X017379.hg.1 |
TC0X001284.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00010525.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171770.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171770.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171770.hg.1 |
ENST00000262839.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00171770.hg.1 |
NM_012471.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00171770.hg.1 |
TC0X00010525.hg.1 |
Clariom D TC |